Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHLHE40 Mouse anti-Human, Clone: OTI4C4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527259

Catalog No. PIMA527259


Add to cart

Description

Description

Differentiated embryo-chondrocyte expressed gene 1 (DEC1) is an important protein involved in embryonic cell differentiation, proliferation, and apoptosis. It has also recently been shown to be regulated by hypoxia. The loss of DEC1 expression may be an early event in the development of lung cancer, while DEC1 may be up-regulated by hypoxia in other cancers and in more extreme hypoxia it may have a role in cell death.
Specifications

Specifications

BHLHE40
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O14503
BHLHB2, DEC1, FLJ99214, HLHB2, SHARP-2, STRA13, Stra14, basic helix-loop-helix domain containing, class B, 2; class B basic helix-loop-helix protein 2; class E basic helix-loop-helix protein 40; differentially expressed in chondrocytes 1; differentially expressed in chondrocytes protein 1; differentiated embryo chondrocyte expressed gene 1; enhancer-of-split and hairy-related protein 2; stimulated by retinoic acid gene 13 protein
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8553
Western Blot
OTI4C4
Unconjugated
BHLHE40
Liquid
BHLHE40
Human recombit protein fragment corresponding to amino acids 1-313 of BHLHE40 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit