Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHBG Mouse anti-Human, Clone: SPM605, Abnova™

Mouse monoclonal antibody raised against full length recombinant human SHBG.

Manufacturer:  Abnova Corporation MAB13155

Catalog No. 89-934-928


Add to cart

Description

Description

This gene encodes a steroid binding protein that was first described as a plasma protein secreted by the liver but is now thought to participate in the regulation of steroid responses. The encoded protein binds each steroid molecule as a dimer formed from identical or nearly identical monomers. The use of alternate promoters and alternatively spliced transcripts have been described. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Specifications

SHBG
SPM605
Mouse monoclonal antibody raised against full length recombinant human SHBG.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
ABP/MGC126834/MGC138391/TEBG
Mouse
IgG1
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
SHBG
Recombinant protein corresponding to full length human SHBG.
Protein A/G purification
RUO
Primary
6462
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit