Learn More
Aviva Systems Biology Corp SHC1 Antibody - C-terminal region (ARP50490_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50490_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SHC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SHC1. Alternate names: FLJ26504, SHC, SHCA, p52SHC, p66, p66SHCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60354