Learn More
Aviva Systems Biology Corp SHC3 Antibody - middle region (ARP56958_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56958_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SHC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SHC3. Alternate names: N-Shc, NSHC, SHCC, RAIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62043