Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHC3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550137

Catalog No. PA550137


Add to cart

Description

Description

The antibody detects endogenous levels of total SHC3 protein.

Signaling adapter that couples activated growth factor receptors to signaling pathway in neurons. Involved in the signal transduction pathways of neurotrophin-activated Trk receptors in cortical neurons.
Specifications

Specifications

SHC3
Polyclonal
Unconjugated
SHC3
Liquid
SHC3
Synthesized peptide derived from internal of human SHC3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q92529, Q61120
NSHC, SHCC
Rabbit
IgG
100 μL
-20°C
Primary
20418, 53358
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit