Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549554

Catalog No. PA549554

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Signaling adapter that couples activated growth factor receptors to signaling pathway in neurons. Involved in the signal transduction pathways of neurotrophin-activated Trk receptors in cortical neurons.
Specifications

Specifications

SHC3
Polyclonal
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q92529, Q61120, O70143
SHC-transforming protein 3, Neuronal Shc, N-Shc, Protein Rai, SHC-transforming protein C, Src homology 2 domain-containing-transforming protein C3, SH2 domain protein C3, SHC3, NSHC, SHCC
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
114858, 20418, 53358
Immunohistochemistry, Western Blot
Unconjugated
SHC3
Liquid
SHC3
synthetic peptide corresponding to 305-330aa of human SHC3
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit