Learn More
CLAD (3), Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against CLAD (3).
Manufacturer:  Abnova Corporation PAB8239
Catalog No. 89-138-658
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
CLAD (3)
Unconjugated
Enzyme Immunoassay (0.312 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
CLAD (3) conjugated with BTG.
RUO
Primary
Not Provided
ELISA
Sheep polyclonal antibody raised against CLAD (3).
In 20mM phosphate buffer, 150mM NaCl, pH 7.2 (0.09% sodium azide)
Sheep
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal