Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyp27b1, Sheep, Polyclonal Antibody, Abnova™

Sheep polyclonal antibody raised against synthetic peptide of Cyp27b1.

Manufacturer:  Abnova Corporation PAB13532

Catalog No. 89-116-727


Add to cart

Description

Description

Sequence: RHVELREGEAAMRNQGKPEEDMPS
Specifications

Specifications

Cyp27b1
Unconjugated
Western Blot (1:500) Immunohistochemistry (1:500) The optimal working dilution should be determined by the end user.
Cyp27b1
Liquid
Cyp27b1
A synthetic peptide corresponding to amino acids 266-289 of mouse Cyp27b1.
RUO
Primary
13115
Human, Murine
Immunohistochemistry, Western Blot
Sheep polyclonal antibody raised against synthetic peptide of Cyp27b1.
In glycine buffered NaCl, 1mM EDTA, pH 7.4 (0.09% sodium azide, 0.1% Epsilon-amino-n-caproic acid, 0.01% benzamidine)
O35084
Cp2b|Cyp1|Cyp27b|Cyp40|MGC91184|P450c1|Pddr|Vdd1|Vddr|VddrI|Vdr
Sheep
100 uL
Store at 4°C. Do not freeze.
Polyclonal
This antibody reacts with Cyp27b1, 56 KDa Protein in Western Blot.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only