Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-FGF basic/FGF2, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236717

Catalog No. NBP236717


Add to cart

Description

Description

FGF basic/FGF2 Polyclonal antibody specifically detects FGF basic/FGF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

FGF basic/FGF2
LYOPH
Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000-1:2000, Immunohistochemistry-Frozen 1:1000-1:2000
P09038
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2
Sheep
IgG
100 ul
Angiogenesis, Cancer
Primary
2247
Reconstitute with 0.1 ml sterilized water.
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Unconjugated
Whole antisera with No Preservative
Antisera
FGF2
Recombinant human basic FGF
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
A high level of specifity for bFGF was shown by immunohistochemistry for this antiserum.
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit