Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-GPT, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244112

Catalog No. NBP244112


Add to cart

Description

Description

Specifically detects GPT in Porcine samples, and it is validated for ELISA, Western Blotting

GPT Polyclonal antibody specifically detects GPT in Porcine samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

GPT
LYOPH
Western Blot 1:500 - 1:3000, ELISA 1:5000 - 1:25000
Purified
GPT
GPT (ALT) [Pig Heart]. Uniprot ID P13191
IgG purified
RUO
Primary
2875
Porcine
Western Blot, ELISA
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
AAT1alanine aminotransferase 1, ALT1, ALT1EC 2.6.1.2, Glutamate pyruvate transaminase 1, glutamic-alanine transaminase 1, Glutamic--alanine transaminase 1, glutamic-pyruvate transaminase (alanine aminotransferase), Glutamic--pyruvic transaminase 1, GPT1GPT 1
Sheep
IgG
0.1 mg
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Reconstitute with 100 ul deionized water.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only