Learn More
Herpes Simplex Virus Type-2, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against Herpes Simplex Virus Type-2.
Manufacturer:  Abnova Corporation PAB13979
Catalog No. 89-137-776
Add to cart
Description
Description
Specifications
Specifications
Herpes Simplex Virus Type-2
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Native from human Herpes Simplex Virus type 2 Strain G.
RUO
Primary
ICPs + late structural (virion) antigens. Does not react with HEp-2 cells. HSV Type-1 titer is 1:512 by indirect immunofluorescence.
ELISA, Immunoblot, Immunofluorescence, Immunoprecipitation, Western Blot
Sheep polyclonal antibody raised against Herpes Simplex Virus Type-2.
In 10mM PBS, pH 7.2 (0.09% sodium azide)
Sheep
1 mL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Herpes Simplex Virus