Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-LRPAP, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP1949811.0 MG

Catalog No. NBP19498110


Add to cart

Description

Description

LRPAP Polyclonal antibody specifically detects LRPAP in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

LRPAP
10.0 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
P30533
A2MRAPMRAP, A2RAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, Alpha-2-MRAP, HBP44, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein associated protein 1, Low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-related protein-associated protein 1(alpha-2-macroglobulin receptor-associated protein 1), MGC138272, RAP
Sheep
IgG
1 mg
Store at -80C. Avoid freeze-thaw cycles.
Polyclonal
Human
ELISA, Western Blot
Unconjugated
50mM Sodium Posphate (pH 6.6), 0.1M NaCl and 1mM EDTA with No Preservative
Purified
LRPAP1
Recombinant full length wild-type human protein (Non-glycosylated).
IgG purified
RUO
Primary
4043
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit