Learn More
Meperidine Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against Meperidine.
Manufacturer:  Abnova Corporation PAB8303
Catalog No. 89-138-721
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Specifications
Specifications
Meperidine
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Meperidine conjugated with BTG.
RUO
Primary
ELISA, Western Blot
Sheep polyclonal antibody raised against Meperidine.
In 20mM phosphate buffer, 150mM NaCl, pH 7.2 (0.09% sodium azide)
Sheep
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal