Learn More
Sheep anti-Prostaglandin F2 alpha, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP1191750.05ML
Catalog No. NBP11917505
Add to cart
Description
Description
Prostaglandin F2 alpha Polyclonal antibody specifically detects Prostaglandin F2 alpha in All Species samples. It is validated for Enzyme Immunoassay.
Specifications
Specifications
Prostaglandin F2 alpha
Unconjugated
Whole antisera with Sodium Azide
PG F2 alpha, PGF 2 alpha, PGF2 alpha, PGF2alpha, Prostaglandin F2alpha, ProstaglandinF2 alpha
8-iso-Prostaglandin F2alpha conjugated to porcine thyroglobulin
Unpurified
RUO
Primary
8-iso-Prostaglandin F2 alpha
ELISA
Enzyme Immunoassay
Antisera
Sheep
IgG
0.05 ml
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
All Species