Learn More
Sheep anti-Prothrombin, Polyclonal, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP1949401.0 MG
Catalog No. NBP19494010
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Prothrombin
10.0 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
P19221
Coagulation factor II, coagulation factor II (thrombin), EC 3.4.21, EC 3.4.21.5, prothrombin, prothrombin B-chain, PT, serine protease
Sheep
IgG
1 mg
Store at -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
ELISA, Western Blot
Unconjugated
50mM Sodium Posphate (pH 6.6), 0.1M NaCl and 1mM EDTA with No Preservative
Purified
F2
Full length native protein (purified) (Mouse).
IgG purified
RUO
Primary
2147