Learn More
SR beta, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against synthetic peptide of SR beta.
Manufacturer:  Abnova Corporation PAB10406
Catalog No. 89-136-971
Add to cart
Description
Description
Specifications
Specifications
SR beta
Unconjugated
Western blot (1 to 5 mg/mL) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to amino acids 246-265 of SR beta.
RUO
Primary
Canine
Immunoprecipitation, Western Blot
Sheep polyclonal antibody raised against synthetic peptide of SR beta.
In PBS (0.08% sodium azide)
Sheep
250 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal