Learn More
Tobramycin, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against Tobramycin.
Manufacturer:  Abnova Corporation PAB8386
Catalog No. 89-138-803
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Tobramycin
Unconjugated
Enzyme Immunoassay (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Tobramycin conjugated with BSA.
RUO
Primary
ELISA
Sheep polyclonal antibody raised against Tobramycin.
In 20mM phosphate buffer, 150mM NaCl, pH 7.2 (0.09% sodium azide)
Sheep
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal