Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Sheep anti-u-Plasminogen Activator/Urokinase, HRP, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP1949291.0 MG

Catalog No. NBP19492910



Description

Description

u-Plasminogen Activator/Urokinase Polyclonal antibody specifically detects u-Plasminogen Activator/Urokinase in Human samples. It is validated for ELISA.
Specifications

Specifications

u-Plasminogen Activator/Urokinase
2.0 mg/mL
ELISA 1:100-1:2000
P00749
ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, uPA, u-PA, U-plasminogen activator, urinary, urokinase-type plasminogen activator
Sheep
IgG
1 mg
Store at -80C. Avoid freeze-thaw cycles.
Polyclonal
Human
ELISA
HRP
PBS (pH 7.4) and 1.0% BSA with 0.01% Thimerosal
Purified
PLAU
Recombinant full length wild-type human protein (Glycosylated).
IgG purified
RUO
Primary
5328
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit