Learn More
Fisher Science Education™ Preserved Specimens: Sheep Brain, Fully Extracted Products for Science Education
Our highest-quality specimen.
Specifications
Preservative Bio-Fresh™
Product Type Sheep Brain
View More Specs
Products 1
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
S1494S10 Products for Science Education
View Documents
10 Pack This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
The dura mater and hypophysis are left intact and undamaged.
INJECTION KEY
  • Plain: Circulatory system not injected with colors
  • Single: Arteries injected with red latex
  • Double: Arteries red, veins blue
  • Triple: Arteries red, veins blue, and hepatic portal veins yellow
Order Info
Packed in Fisher Science Education™ Bio-Fresh™.
Specifications
Specifications
Bio-Fresh™
Sheep Brain