Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Sheep Clusterin/APOJ ELISA Kit (Colorimetric)

Sheep Clusterin/APOJ ELISA Kit (Colorimetric) employs Sandiwch ELISA technique for detection of Sheep Clusterin/APOJ

$602.00

Specifications

Species Sheep
Sensitivity 4.963ng/mL
Sample Type serum, plasma and tissue homogenates
Research Discipline Cardiovascular Biology
Synonym 40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin (complement lysis inhibitor, SP-40, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2TRPM2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB599921KT View Documents Novus Biologicals™
NBP2599921KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Sheep Clusterin/APOJ ELISA Kit (Colorimetric) measures sheep Clusterin/APOJ in serum, plasma and tissue homogenates.
Specifications

Specifications

Sheep
serum, plasma and tissue homogenates
40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin (complement lysis inhibitor, SP-40, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2TRPM2
Clusterin/APOJ
1191
ELISA Kit (Colorimetric)
50 - 100μL
4.963ng/mL
Cardiovascular Biology
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
ELISA
CLU
7.813ng/mL to 500ng/mL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit