Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep IgG, Fc, Biotin, Isotype control, Novus Biologicals™
SDP

Sheep Antibody

Supplier:  Novus Biologicals NBP197087

Catalog No. NBP197087


Add to Cart

Description

Description

IgG Fc specifically detects IgG Fc in Sheep samples. It is validated for Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications

Specifications

IgG Fc
Biotin
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Multi-step
RUO
No reaction was observed against anti-Sheep IgG F(ab')2 or anti- Papain.
Sheep
IgG
LYOPH
Restore with 1.0mL of deionized water (or equivalent).
Sheep
1 mg
Primary
Reconstitute with 1.0 ml deionized water (or equivalent).
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit