Learn More
Sheep IgG, Fc, Fluorescein, Isotype control, Novus Biologicals™
Sheep Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196986
Catalog No. NBP196986
Add to cart
Description
Description
IgG Fc antibody specifically detects IgG Fc in Sheep samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
IgG Fc
LYOPH
Immunocytochemistry/Immunofluorescence 1:500 - 1:2500
Conjugated
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Sheep IgG, anti-Sheep IgG F(c) and anti-Sheep Serum. No reaction was observed against anti-Sheep IgG F(ab)2 or anti-Papain.
Sheep
Immunocytochemistry, Immunofluorescence
Fluorescein
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
Sheep
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.