Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Novus Biologicals™ Sheep Progesterone ELISA Kit (Colorimetric)

Sheep Progesterone ELISA Kit (Colorimetric) employs competitive ELISA technique for detection of Sheep Progesterone

$602.00

Specifications

Species Sheep
Sensitivity 0.2ng/mL
Sample Type serum, plasma, tissue homogenates
Research Discipline Signal Transduction
Synonym Pregn 4 ene 3 20 dione
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601211KT View Documents Novus Biologicals™
NBP2601211KIT
1 Kit Each for $602.00
N/A
Description

Description

The Sheep Progesterone ELISA Kit (Colorimetric) measures sheep Progesterone in serum, plasma, tissue homogenates.
Specifications

Specifications

Sheep
serum, plasma, tissue homogenates
Pregn 4 ene 3 20 dione
Progesterone
ELISA Kit (Colorimetric)
50 - 100μL
0.2ng/mL
Signal Transduction
Store at 4°C
ELISA
0.4ng/mL to 30ng/mL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit