Learn More
Zyagen LabsSupplier Diversity Partner Sheep universal reference cDNA is a full-length first strand cDNA synthesized from Uni reference RNA isolated from wide variety of male and female sheep tissues. Ideal for cloning, sequencing and as a control for qPCR. 40 reactions/PK DFS Item
Manufacturer:  Zyagen LabsSupplier Diversity Partner SDUR40
Zyagen sheep Universal Reference cDNA is a full length first strand cDNA synthesized from Zyagen universal reference RNA which is isolated from wide variety of male and female tissues. The cDNA is primed with oligo dT and can be used in gene cloning, target sequencing and as a control for gene expression analysis by regular PCR and real time PCR. Each cDNA product is enough for 40 PCR reactions. Features: Synthesized from high quality intact universal reference total RNA. Consistent control for standard data set comparisons. Prepared from several major tissues for broad gene coverage. Made at industrial scales to minimize lot-to-lot variation. Quality Control: The integrity of each RNA sample used for cDNA synthesis is verified by denatured agarose gel electrophoresis. The purity of RNA is assessed by NanoDrop (A260/A280: 1.9-2.1). Residual DNA contamination is tested by PCR. The synthesized cDNA is also used as template PCR amplification of ß-actin gene.
Catalog No. 50-171-8097