Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Sheep Rotavirus Antigen ELISA Kit (Colorimetric)
Assay has high sensitivity and specificity for detection of sheep Rotavirus antigen
Supplier: Novus Biologicals™ NBP2601341KIT
Description
The Sheep Rotavirus Antigen ELISA Kit (Colorimetric) measures Sheep Rotavirus Antigen in feces.Specifications
Qualitative | |
Fecal Extract | |
Sheep | |
Assay plate, Adhesive Strip (For 96 wells), HRP-conjugate, Wash Buffer, Substrate A, Substrate B, Stop Solution, Instruction manual, Positive Control, Negative Control | |
Cell Biology | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. |
ELISA Kit (Colorimetric) | |
For detection of sheep Rotavirus antigen | |
ELISA | |
1 Kit | |
50 to 100 μL | |
Rotavirus Antigen |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction