Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Sheep Rotavirus Antigen ELISA Kit (Colorimetric)

Assay has high sensitivity and specificity for detection of sheep Rotavirus antigen

$602.00

Specifications

Species Sheep
Sample Type feces
Research Discipline Cell Biology
Specificity For detection of sheep Rotavirus antigen
Storage Requirements Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601341KT View Documents Novus Biologicals™
NBP2601341KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Sheep Rotavirus Antigen ELISA Kit (Colorimetric) measures Sheep Rotavirus Antigen in feces.
Specifications

Specifications

Sheep
Cell Biology
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
ELISA
Qualitative
Assay plate, Adhesive Strip (For 96 wells), HRP-conjugate, Wash Buffer, Substrate A, Substrate B, Stop Solution, Instruction manual, Positive Control, Negative Control
feces
For detection of sheep Rotavirus antigen
Rotavirus Antigen
ELISA Kit (Colorimetric)
50 - 100μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit