Learn More
BioworldSupplier Diversity Partner Sheep Serum, 100 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306110831
All animals are certified of U.S. origin. This is an exceptional quality serum.Specification: Raw, non-sterile Low hemoglobin levels Physiochemical constituents measured to ensure lot to lot consistency Endotoxin testing available Mycoplasma testing available Storage conditions: -20° ± 5°CShips on dry ice.
Catalog No. 50-254-192