Shelby™ FDP™ Leather Gloves
Fire-retardant and heat-resistant heavyweight gloves meet the demands of firefighting
Manufacturer: Shelby Specialty Gloves 5228L
 View more versions of this product
Catalog No. 19-100-349
  Check Availability
Add to Cart
    Specifications
    Certifications NFPA 1971, CE PPE, EN 659:1996, EN 388:1994
    Glove Cuff Style Gauntlet
    Material FDP Leather
    Liner Material Thermal
    Size Large
    Color Midnight blue
  • Description & Specifications
Related Products
Product Content Feedback