Learn More
Shelby™ FDP™ Leather Gloves
Encompass
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Fire-retardant and heat-resistant heavyweight gloves meet the demands of firefighting
  • Durable outer shells are made of either soft, koala-tanned cowhide leather or abrasion-resistant brushed pigskin
  • Gloves feature Gore RT7100 glove barrier fabric laminated to SEF modacrylic fleece
  • Lining is sewn to shell at fingertips and wrist to eliminate pullouts
  • Seams are sealed with DuPont™ silicone sealant
  • Gunn cut with wing thumb construction
  • Welted thumb and fourchette seams
  • Glove body extends 2 in. (5 cm) above wrist crease with shirred elastic at wrist
  • 0.25 in. (6.3 mm) hanger loop for easy storage
Certifications
Manufactured in an ISO 9002 Certified Facility
Compliance
NFPA 1971-2013
Certifications/Compliance NFPA 1971, CE PPE, EN 659:1996, EN 388:1994
Color Tan
Material Cowhide Leather, Pigskin Leather
Product Type Structural Firefighting Gloves
Size Small
Glove Cuff Style Gauntlet
Liner Material Thermal