Shelby™ FDP™ Leather Gloves
Fire-retardant and heat-resistant heavyweight gloves meet the demands of firefighting
Manufacturer: Shelby Specialty Gloves 5225XS
 View more versions of this product
Catalog No. 19-821-482
  Check Availability
Add to Cart
    Specifications
    Certifications NFPA 1971, CE PPE, EN 659:1996, EN 388:1994
    Glove Cuff Style Knit
    Material FDP Leather, Cowhide, Brushed Pig Skin Leather
    Liner Material Thermal
    Size X-Small
    Color Tan
  • Description & Specifications
Related Products
Product Content Feedback