Shelby™ FDP™ Leather Gloves
Fire-retardant and heat-resistant heavyweight gloves meet the demands of firefighting
Manufacturer: Shelby Specialty Gloves 5227-XL
 View all options for this product
Catalog No. 19-821-491
  Check Availability
Add to Cart
    Color Midnight blue
    Glove Cuff Style Knit
    Material FDP Leather, Nomex, Cowhide Leather
    Liner Material Thermal
    Size X-Large
    Certifications NFPA 1971, CE PPE, EN 659:1996, EN 388:1994
    • Durable outer shells are made of either soft, koala-tanned cowhide leather or abrasion-resistant brushed pigskin
    • Utilize Gore RT7100 glove barrier fabric laminated to SEF modacrylic fleece
    • Lining is sewn to shell at fingertips and wrist to eliminate pullouts
    • Seams are sealed with DuPont™ silicone sealant
    • Gunn cut with wing thumb construction; thumb and forchette seams are welted
    • Glove body extends 2 in. (5cm) above wrist crease with shirred elastic at wrist
    • 0.25 in. (6.3mm) hanger loop for easy storage
  • Description & Specifications
Related Products
Product Content Feedback