Learn More
Shelby™ Side Split Leather Gloves with Cotton Flannel Lining
Encompass
Click to view available options
Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Premium side-split leather gloves feature a wing thumb, shirred elastic back, and cotton flannel palm lining.
  • Premium side split leather
  • Shirred elastic back
  • Cotton flannel palm
  • 2.5 in. rubberized safety cuff
Cuff Material Rubberized
Grade Premium
Hand Specific
Material Side Split Cowhide Leather
Product Type Driver Gloves
Size Large
Glove Cuff Style Open
Liner Material Flannel
Series 4142
Thumb Style Wing
Length (English) Cuff 2.5 in.
Length (Metric) Cuff 6.4 cm