Learn More
Shelby™ Proximity Gloves Non-distribution product as customer accommodation.
ARFF glove offers reflective/radiant heat protection
Manufacturer:  Shelby™ 5200XL
 View more versions of this product
Catalog No. 19-821-466
Add to cart
Specifications
Specifications
Special gripping
Steamblock insulative pad
X-Large
Wristlet
PBI™, aluminized, Dupont™ Kevlar™ aramid fibers
NFPA 1976, 2007 edition