Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
Shelby Proximity Gloves
ARFF glove offers reflective/radiant heat protection
$244.50 - $246.00
Catalog Number | Mfr. No. | Material | Size | Glove Cuff Style | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Size | Glove Cuff Style | Price | Quantity & Availability | |||
19-821-464
![]() |
Shelby
5200L |
Dupont™ Kevlar™ Aramid Fiber | Large | Wristlet |
Pair for $244.50
Dozen of 12 PR for $2,934.00 |
N/A |
19-100-343
![]() |
Shelby
5200J |
Split Leather | 2X-Large | Knit |
Pair for $246.00
Dozen of 12 PR for $2,952.00 |
N/A |
19-100-344
![]() |
Shelby
5200S |
Split Leather | Small |
Pair for $244.50
Dozen of 12 PR for $2,934.00 |
N/A |
19-821-466
![]() |
Shelby
5200XL |
Dupont™ Kevlar™ Aramid Fiber | X-Large | Wristlet |
Pair for $244.50
Dozen of 12 PR for $2,934.00 |
N/A |
Description
- Aluminized PBI/Kevlar knit ARFF glove offers reflective/radiant heat protection on the top, sides and in between the fingers
- Steamblock insulative pad on back of hand
- Special gripping surfaces applied to the palm
Compliance
Meet NFPA
Specifications
NFPA 1976, 2007 edition |