Learn More
Shelby™ Specialty 5280 Gloves
Encompass
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Specialty thermal gloves are resistant to flames, heat, and punctures for fire management or other industrial applications.
  • Heavyweight, gauntlet-style gloves
  • Grain elkhide leather along with Shelby abrasion-resistant brushed pigskin leather
  • Shelby Black Line Nomex™ 101.6 mm (4 in.) wristlet
  • GORE RT7100 glove barrier fabric
  • Gunn-cut pattern with wing thumb and wrap-around index finger
Certifications
SEI Certified
Certifications/Compliance NFPA 1971, CE PPE, EN 659:1996, EN 388:1994
Color Tan
Material Pigskin Leather, Elkskin Leather
Product Type Structural Firefighting Gloves
Size X-Large
Glove Cuff Style Gauntlet
Liner Material Thermal