Learn More
Shelby™ Xtrication™ Gloves Non-distribution product as customer accommodation.
Anatomically designed for improved hand and finger dexterity
Manufacturer:  Shelby™ 2500S
 View more versions of this product
Catalog No. 19-500-482
Add to cart
Description
Description
  • Glove body constructed of durable nylon Cordura™
  • Shirred elastic wrist closure
  • Palm and finger reinforcements cover all high-wear stress points with Kevlar™ fabric
  • Rubberized fingers enhance grip in wet or slippery situations
  • Black with chartreuse glove back for improved visibility
Specifications
Specifications
Safety
Fluffed Terry Cotton
Black
Cordura™ Nylon
Small
CE PPE, EN 659:1996, EN 388:1994