Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHF Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548899

Catalog No. PA548899

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Adapter protein which may play a role in the regulation of apoptosis in response to PDGF.
Specifications

Specifications

SHF
Polyclonal
PBS with 0.09% sodium azide
Q7M4L6
SH2 domain-containing adapter protein F, SHF
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
90525
Western Blot
Unconjugated
SHF
Liquid
SHF
KLH conjugated synthetic peptide between 384-411 amino acids from the C-terminal region of human SHF.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit