Learn More
Fisherbrand Shield Face Shield
Reduce particle generation
Specifications
Color | Blue |
---|---|
Construction | Thin middle layer of Meltblown Polypropylene filter media lies between Spunbond Polypropylene inner and outer layers ultrasonically welded to reduce particle generation |
Material | Polypropylene |
Catalog Number | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Includes | Price | Quantity & Availability | ||||||
18-960A
|
Ear loops and clips |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
18-960B
|
Ear loops |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
Description
Thin middle layer of meltblown polypropylene filter media lies between spunbonded polypropylene inner and outer layers.
- Bacterial filtration efficiency is 99%
- Layers ultrasonically welded to reduce particle generation
- Blue outer layer with white inner layer
- Elastic ear loops or polypropylene ties, with or without plastic clip
Specifications
Blue | |
Thin middle layer of Meltblown Polypropylene filter media lies between Spunbond Polypropylene inner and outer layers ultrasonically welded to reduce particle generation | |
Polypropylene |