Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shigella dysenteriae Serotype 10 Monoclonal Antibody (1006/91), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MA546609

Encompass

Catalog No. PIMA546609


Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

Description

Description

Shigella dysenteriae Serotype 10 Monoclonal antibody specifically detects Shigella dysenteriae Serotype 10 in Bacteria samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Shigella dysenteriae Serotype 10
Monoclonal
3.4 mg/mL
PBS with 0.1% sodium azide
Mouse
Protein A/G
RUO
Bacteria
Liquid
ELISA
1006/91
Unconjugated
S. dysenteriae
S. dysenteriae, NCTC 9351
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG3
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit