Learn More
Seracare Life Sciences IncSupplier Diversity Partner SHIGELLA SPECIES POS CTRL  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 53700003
Heat-killed Shigella boydii, Shigella flexneri, and Shigella dysenteriae cells in dextran solution. Lot specific cell counts can be obtained by requesting a Certificate of Analysis or by calling Technical Services with the specific lot number.
Catalog No. 50-674-58
Due to product restrictions, please to purchase or view availability for this product.