Learn More
Trajan™ Shimadzu Instrument Ferrules
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
$84.50 - $98.70
Specifications
Type Ferrule
View More Specs
Products 3
Catalog Number Mfr. No. For Use With (Equipment) Material Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Material Price Quantity & Availability  
SG073204
View Documents
Trajan™
073204
QP5000/5050 Standard MS Interface SilTite MetalFor 0.10-0.25 mm ID Columns 2 nuts included
Pack of 10 for $98.70
Add to cart
SG073227
View Documents
Trajan™
073227
SGE; GC14 A, GC17 A, GC2010 and GC2014 Detector/Injectors SilTite Metal, initial installation 10 ferrules, 2 nuts; Column size: 0.10 to 0.25 mm ID columns
Pack of 10 for $98.70
Add to cart
SG0726564
View Documents
Trajan™
0726564
QP5000/5050 Standard MS Interface 15% Graphite, 85% Vespel ferrules; Column size: QP5000-I 0.32 mm ID columns
Pack of 10 for $84.50
Add to cart
Description
Description
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
Ferrule