Learn More
SHIP-1 Mouse, PE, Clone: 32/SHIP-1, BD Pharmingen™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566009
Catalog No. BDB566009
Add to cart
Description
Description
SHIP-1 Monoclonal antibody specifically detects SHIP-1 in Mouse, Rat samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
Specifications
SHIP-1
32/SHIP-1
PE
Purified
Mouse
Mouse IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed.
Monoclonal
Flow Cytometry, Flow Cytometry (Intracellular Staining)
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Ship1; Ship; Inpp5d; p150Ship; SIP-145; s-SHIP; 7a33
Mouse p150 [SHIP] aa. 16-135
50μg
Cell Biology
Primary
Murine, Rat
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.