Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHIP Mouse anti-Human, Janelia Fluor 549, Clone: SHIP-02, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP145077JF549
Description
SHIP Monoclonal specifically detects SHIP in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
SHIP | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
INPP5D | |
Peptide coresponding to a sequence within N-terminal domain of Human SHIP-1. | |
0.1 mL | |
Primary | |
The antibody SHIP-02 reacts with SHIP-1, a phosphoinositide phosphatase largely confined to hematopoietic cells. Multiple forms of SHIP-1 have been reported with molecular weights of 110, 125, 130, 135 and 145 kDa. | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry | |
SHIP-02 | |
Flow Cytometry | |
145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, MGC104855, SHIPMGC142140 | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Immunology, Signal Transduction | |
3635 | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title