Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SHIP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583381
Description
Immunogen sequence: TLGELIGLYA QPNQGLVCAL LLPVEGEREP DPPDDRDASD GEDEKPPLPP RSGSTSISAP TGPSSPLPAP ETPTAPAAES APNGLST.
The protein encoded by this gene is an SH2-containing 5'-inositol phosphatase that is involved in the regulation of insulin function. The encoded protein also plays a role in the regulation of epidermal growth factor receptor turnover and actin remodeling. Additionally, this gene supports metastatic growth in breast cancer and is a valuable biomarker for breast cancer.
Specifications
SHIP2 | |
Polyclonal | |
Unconjugated | |
INPPL1 | |
51C; 51C protein; ablSH3-binding protein; inositol polyphosphate phosphatase like 1; inositol polyphosphate phosphatase-like 1; inositol polyphosphate phosphatase-like protein 1; Inppl1; INPPL-1; OPSMD; Phosphatidylinositol 3,4,5-trisphosphate 5-phosphatase 2; phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 2; protein 51C; SH2 domain-containing inositol 5'-phosphatase 2; SH2 domain-containing inositol phosphatase 2; SH2 domain-containing inositol-5'-phosphatase 2; SH2-containing inositol phosphatase 2; SHIP2; SHIP-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3636 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15357 | |
INPPL1 | |
Recombinant protein corresponding to Human INPPL1. Recombinant protein control fragment (Product #RP-96522). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction