Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHISA9 isoform X2 Rabbit anti-Human, Mouse, Non-human Primate, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc CKAMP44AB112AP
Description
SHISA9 isoform X2 Polyclonal antibody specifically detects SHISA9 isoform X2 in Human, Mouse, Non-human Primate samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
SHISA9 isoform X2 | |
Polyclonal | |
Unconjugated | |
Shisa9 | |
2700045P11Rik; FLJ10015; SHISA9 | |
Synthetic peptide taken within amino acid region 300-400 on rat CKAMP44A/B protein. | |
200 μL | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
B4DS77, Q9CZN4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
72555, 729993, 739962 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction