Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHMT2 Mouse anti-Human, Clone: OTI1E6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene CF808814
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Mammalian serine hydroxymethyltransferase (SHMT) is a tetrameric, pyridoxal phosphate (PLP)-dependent enzyme that catalyzes the reversible interconversion of serine and tetrahydrofolate to glycine and methylenetetrahydrofolate in the cytoplasm (cSHMT, SHMT1) and mitochondria (mSHMT, SHMT2). cSHMT preferentially supplies one-carbon units for thymidylate biosynthesis, depletes methylenetetrahydrofolate pools for S-adenosylmethionine (SAM) synthesis by synthesizing serine, sequesters 5-methyltetrahydrofolate, and inhibits SAM synthesis.Specifications
SHMT2 | |
Monoclonal | |
Unconjugated | |
SHMT2 | |
GLYA, HEL-S-51e, SHMT | |
Mouse | |
Affinity Chromatography | |
RUO | |
6472 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin) | |
OTI1E6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P34897 | |
SHMT2 | |
Full length human recombit protein of human SHMT2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction