Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synoptics™ Short Pass Filter with Screw Thread
GREENER_CHOICESDP

Short pass filter for use in Syngene Ingenius System

Supplier:  Synoptics™ FILTSP58

Catalog No. 01-257-211


Explore available promotions
Add to Cart

Description

Description

  • For use in Syngene Ingenius System
  • 58mm filter
Specifications

Specifications

Fixed
Filter Short Pass
Ingenius 3, Nugenius, Nugenius+
515 to 599 nm
GeneGenius, MultiGenius, ChemiGenius
58 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.