Learn More
Showa™ 377 Dipped Nitrile Gloves
Featuring a liquid proof nitrile undercoat for oil protection and an additional foam nitrile coating on top for excellent grip in wet and oily applications. SHOWA™ 377 is a double dipped nitrile glove that provides added protection from oils and fl
$754.35
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-020-701
View Documents
Showa™
377XXL10
X-Large Case of 72 for $754.35
Add to cart
19-020-700
View Documents
Showa™
377S06
Small Case of 72 for $754.35
Add to cart
Description
Description
  • Fully coated, extra coating on palm
  • Reinforced knit cuff, scalloped edge
  • Standard finish
  • Without special treatment
  • Anatomical shape
  • Seamless knit
  • A flexible, robust glove offering great dexterity and with good resistance to tearing
  • Glove stays correctly adjusted
  • Protects the hand from oils, hydrocarbons, grease and abrasion, with optimal long lasting grip
  • Impermeable for working in damp or greasy environments, with increased resistance to abrasion for better durability
  • Minimal allergy risks
  • Designed for easy movement and continuous wear
  • No irritation by seams in contact with the skin
  • Wrist well protected