Learn More
SHOWA™ 4560 Series Gloves
Crafted with high performance, chemical resistant Zorb-It Technology. SHOWA™ 4560 is equipped with Aramid fibers for first class cut protection and a long lasting grip.
Specifications
Certifications/Compliance ISO 13997
Color Yellow
Cut Resistance Rating 3
Description Nitrile Coating
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-020-707
View Documents
SHOWA™
456011
2X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-020-702
View Documents
SHOWA™
456006
X-Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • ANSI cut level A3
  • ISO 13997 cut level C
  • Cut-resistant liner
  • Ergonomic design for maximum comfort
  • Unparalleled grip
  • Sponge nitrile technology absorbs oil and water
  • 15-gauge seamless stretch, cut-resistant liner
  • Made with Kevlar™ aramid fibers
  • Color coded cuff
  • Maximum Oil Absorbency Sponge nitrile coated palm
Specifications
Specifications
ISO 13997
Yellow
3
Nitrile Coating