Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ AEGIS HP54™ Cut-Resistant Gloves Available on GSA/VA Contract for Federal Government customers only.

$288.50

Specifications

Coating Material Sponge Nitrile
Color Black, White
Cut Resistance Rating Level 4
Material HPPE, Lycra™, Nylon
For Use With (Application) Oily surfaces, Metal stamping, mechanical and automotive, sheet Metal assembly, Glass handling, bottling, recycling
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-120-551 Available on GSA/VA Contract for Federal Government customers only.
View Documents
SHOWA™
23007
Small
Dozen of 12 for $288.50
Case of 6 DZ for $1,376.00
Add to cart
 
19-120-553 Available on GSA/VA Contract for Federal Government customers only.
View Documents
SHOWA™
23009
Large
Dozen of 12 for $288.50
Case of 6 DZ for $1,376.00
Add to cart
 
19-120-552 Available on GSA/VA Contract for Federal Government customers only.
View Documents
SHOWA™
23008
Medium
Dozen of 12 for $288.50
Case of 6 DZ for $1,376.00
Add to cart
 
19-120-554 Available on GSA/VA Contract for Federal Government customers only.
View Documents
SHOWA™
23010
X-Large
Dozen of 12 for $288.50
Case of 6 DZ for $1,376.00
Add to cart
 
Description

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Sponge Nitrile
Level 4
Oily surfaces, Metal stamping, mechanical and automotive, sheet Metal assembly, Glass handling, bottling, recycling
Black, White
HPPE, Lycra™, Nylon
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit